Learn More
Biomatik Corporation ADAM10 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07337
Encompass_Preferred
The ADAM10 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Disintegrin and metalloproteinase domain-containing protein 10 protein (214-504AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Disintegrin and metalloproteinase domain-containing protein 10 (EC:3.4.24.81), ADAM 10, ADAM10, KUZ, MADM, CDw156, Kuzbanian protein homolog, Mammalian disintegrin-metalloprotease, CD_antigen: CD156c. UniProt: O14672. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6923