Learn More
Biomatik Corporation PSMD11 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506929
Encompass_Preferred
Psmd11 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PSMD11. The Psmd11 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 26S Proteasome Non-Atpase Regulatory Subunit 11 Protein (2-422Aa). Applications: ELISA, IHC, IF. Target Synonyms: PSMD11. Accession Number: O00231. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6929 Supplier Biomatik Corporation Supplier No. CAC07343
Psmd11 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PSMD11. The Psmd11 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 26S Proteasome Non-Atpase Regulatory Subunit 11 Protein (2-422Aa). Applications: ELISA, IHC, IF. Target Synonyms: PSMD11. Accession Number: O00231. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.