Learn More
Biomatik Corporation DAOA anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07346
Encompass_Preferred
The DAOA Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human D-amino acid oxidase activator protein (1-153AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: Protein G72 DAOA G72. UniProt: P59103. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6932