Learn More
Biomatik Corporation DAPK2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07347
Encompass_Preferred
The DAPK2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Death-associated protein kinase 2 protein (222-370AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: DAP kinase 2, DAP-kinase-related protein 1, DRP-1, DAPK2. UniProt: Q9UIK4. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6933