Learn More
Biomatik Corporation B4GALT3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07348
Encompass_Preferred
The B4GALT3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Beta-1,4-galactosyltransferase 3 protein (34-393AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: UDP-Gal:beta-GlcNAc beta-1, 4-galactosyltransferase 3 UDP-galactose:beta-N-acetylglucosamine beta-1, 4-galactosyltransferase 3. UniProt: O60512. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6934