Learn More
Biomatik Corporation DRD5 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07349
Encompass_Preferred
The DRD5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human D (1B) dopamine receptor protein (361-477AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: D (1B) dopamine receptor, D (5) dopamine receptor, D1beta dopamine receptor, Dopamine D5 receptor, DRD5, DRD1B, DRD1L2. UniProt: P21918. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6935