Learn More
Biomatik Corporation G0S2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07352
Encompass_Preferred
The G0S2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human G0/G1 switch protein 2 protein (1-103AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200. Alternative Names: G0/G1 switch protein 2, G0/G1 switch regulatory protein 2, Putative lymphocyte G0/G1 switch gene, G0S2. UniProt: P27469. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6938