Learn More
Biomatik Corporation IFIT3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07359
Encompass_Preferred
Ifit3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IFIT3. The Ifit3 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interferon-Induced Protein With Tetratricopeptide Repeats 3 Protein (1-490Aa). Applications: ELISA, WB, IHC, IF, IP. Target Synonyms: IFIT3. Accession Number: O14879. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6945