Learn More
Biomatik Corporation HMGB1 Monoclonal Antibody
Supplier:  Biomatik Corporation CAF50662
Encompass_Preferred
HMGB1 Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to High Mobility Group Protein B1. The HMGB1 Monoclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Mouse. Species: Human (Homo sapiens). Immunogen: 1~215aa. Applications: ELISA, WB, IHC, IF. Target Synonyms: High Mobility Group Protein B1. Accession Number: P09429. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9861