Learn More
Biomatik Corporation GPX4 Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50549
Encompass_Preferred
GPX4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Glutathione Peroxidase 4 (Phospholipid Hydroperoxidase). The GPX4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: 30~197aa. Applications: ELISA, WB, IHC, IF. Target Synonyms: Glutathione Peroxidase 4 (Phospholipid Hydroperoxidase). Accession Number: P36969. Gene ID: 2879. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9748