Learn More
Biomatik Corporation HSP70 Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50556
Encompass_Preferred
HSP70 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Heat Shock 70Kda Protein 1A. The HSP70 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, WB, IF, FC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: 291~641aa. Applications: ELISA, IHC, WB, IF, FC, IP. Target Synonyms: Heat Shock 70Kda Protein 1A. Accession Number: P0DMV8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9755