Learn More
Biomatik Corporation ERK1/2 Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50535
Encompass_Preferred
ERK1/2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Mitogen-Activated Protein Kinase 1. The ERK1/2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Please Inquire. Applications: ELISA, WB, IHC, IF. Target Synonyms: Mitogen-Activated Protein Kinase 1. Accession Number: P28482. Gene ID: 5594. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9734