Learn More
Biomatik Corporation SDHB Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50603
Encompass_Preferred
SDHB Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Succinate Dehydrogenase Complex, Subunit B, Iron Sulfur (Ip). The SDHB Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Please Inquire. Applications: ELISA, WB, IHC, IP. Target Synonyms: Succinate Dehydrogenase Complex, Subunit B, Iron Sulfur (Ip). Accession Number: P21912. Gene ID: 6390. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9802