Learn More
Biomatik Corporation Pepsin (Pp) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29795
Encompass_Preferred
Pepsin (Pp) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Pepsin (PP). The Pepsin (Pp) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Val63~Phe163 + Gly206~Ser295. Applications: WB, IHC, ICC, IP. Target Synonyms: Pepsin (PP). Accession Number: P0DJD8. Gene ID: 643834. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9610