Learn More
Biomatik Corporation Heat Shock Protein 70 (Hsp70) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU30421
Encompass_Preferred
Heat Shock Protein 70 (Hsp70) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Heat Shock Protein 70 (HSP70). The Heat Shock Protein 70 (Hsp70) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Asp285~Asp641. Applications: WB, IHC, ICC, IP. Target Synonyms: Heat Shock Protein 70 (HSP70). Accession Number: P0DMV8. Gene ID: 3303. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9643