Learn More
Biomatik Corporation Beta Actin (Actb) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU30721
Encompass_Preferred
Beta Actin (Actb) Monoclonal Antibody is a Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus), Dog (Canis familiaris; Canine), Pig (Sus scrofa; Porcine), Bovine (Bos taurus) Monoclonal Antibody to Beta Actin (ACTB). The Beta Actin (Actb) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus), Dog (Canis familiaris; Canine), Pig (Sus scrofa; Porcine), Bovine (Bos taurus). Immunogen: Met1~Phe375. Applications: WB, IHC, ICC, IP. Target Synonyms: Beta Actin (ACTB). Accession Number: P60709. Gene ID: 60. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9650