Learn More
Biomatik Corporation Ubiquitin Cross Reactive Protein (Ucrp) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU21435
Encompass_Preferred
Ubiquitin Cross Reactive Protein (Ucrp) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Ubiquitin Cross Reactive Protein (UCRP). The Ubiquitin Cross Reactive Protein (Ucrp) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Gly2~Gly157. Applications: WB, IHC, ICC, IP. Target Synonyms: Ubiquitin Cross Reactive Protein (UCRP). Accession Number: P05161. Gene ID: 9636. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9206