Learn More
Biomatik Corporation Fidgetin (Fign) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU21795
Encompass_Preferred
Fidgetin (Fign) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Fidgetin (FIGN). The Fidgetin (Fign) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Leu308. Applications: WB, IHC. Target Synonyms: Fidgetin (FIGN). Accession Number: Q5HY92. Gene ID: 55137. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9218