Learn More
Biomatik Corporation Nexilin (Nexn) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU21941
Encompass_Preferred
Nexilin (Nexn) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Nexilin (NEXN). The Nexilin (Nexn) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Ala214~Gln455. Applications: WB, IHC. Target Synonyms: Nexilin (NEXN). Accession Number: Q9Z2J4. Gene ID: 246172. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9232