Learn More
Biomatik Corporation Gremlin 1 (Grem1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU24640
Encompass_Preferred
Gremlin 1 (Grem1) Polyclonal Antibody is a Chicken (Gallus) Polyclonal Antibody to Gremlin 1 (GREM1). The Gremlin 1 (Grem1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Chicken (Gallus). Immunogen: Arg27~Asp184. Applications: WB, IHC, ICC, IP. Target Synonyms: Gremlin 1 (GREM1). Accession Number: O73755. Gene ID: 395826. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9360