Learn More
Biomatik Corporation Steroidogenic Acute Regulatory Protein (Star) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU22219
Encompass_Preferred
Steroidogenic Acute Regulatory Protein (Star) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Steroidogenic Acute Regulatory Protein (STAR). The Steroidogenic Acute Regulatory Protein (Star) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Cys285. Applications: WB, IHC, ICC, IP. Target Synonyms: Steroidogenic Acute Regulatory Protein (STAR). Accession Number: P49675. Gene ID: 6770. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9246