Learn More
Biomatik Corporation Microfibrillar Associated Protein 5 (Mfap5) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU22440
Encompass_Preferred
Microfibrillar Associated Protein 5 (Mfap5) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Microfibrillar Associated Protein 5 (MFAP5). The Microfibrillar Associated Protein 5 (Mfap5) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Gly25~Pro152. Applications: WB, IHC, ICC, IP. Target Synonyms: Microfibrillar Associated Protein 5 (MFAP5). Accession Number: Q9QZJ6. Gene ID: 50530. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9259