Learn More
Biomatik Corporation Cathepsin S (Ctss) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU25044
Encompass_Preferred
Cathepsin S (Ctss) Polyclonal Antibody is a Bovine (Bos taurus) Polyclonal Antibody to Cathepsin S (CTSS). The Cathepsin S (Ctss) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Bovine (Bos taurus). Immunogen: Leu115~Ile331. Applications: WB, IHC, ICC, IP. Target Synonyms: Cathepsin S (CTSS). Accession Number: P25326. Gene ID: 327711. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9376