Learn More
Biomatik Corporation Solute Carrier Family 3, Member 2 (Slc3A2) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU22843
Encompass_Preferred
Solute Carrier Family 3, Member 2 (Slc3A2) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Solute Carrier Family 3, Member 2 (SLC3A2). The Solute Carrier Family 3, Member 2 (Slc3A2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Ile285~Gln521. Applications: WB, IHC, ICC, IP. Target Synonyms: Solute Carrier Family 3, Member 2 (SLC3A2). Accession Number: P10852. Gene ID: 17254. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9274