Learn More
Biomatik Corporation Myosin Heavy Chain 4, Skeletal Muscle (Myh4) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU23357
Encompass_Preferred
Myosin Heavy Chain 4, Skeletal Muscle (Myh4) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Myosin Heavy Chain 4, Skeletal Muscle (MYH4). The Myosin Heavy Chain 4, Skeletal Muscle (Myh4) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Gln1259~Ser1495. Applications: WB, IHC, ICC, IP. Target Synonyms: Myosin Heavy Chain 4, Skeletal Muscle (MYH4). Accession Number: Q5SX39. Gene ID: 17884. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9297