Learn More
Biomatik Corporation Guanylate Binding Protein 4 (Gbp4) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU25338
Encompass_Preferred
Guanylate Binding Protein 4 (Gbp4) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Guanylate Binding Protein 4 (GBP4). The Guanylate Binding Protein 4 (Gbp4) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Lys292. Applications: WB, IHC. Target Synonyms: Guanylate Binding Protein 4 (GBP4). Accession Number: Q96PP9. Gene ID: 115361. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9389