Learn More
Biomatik Corporation Kallikrein 2 (Klk2) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU23565
Encompass_Preferred
Kallikrein 2 (Klk2) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Kallikrein 2 (KLK2). The Kallikrein 2 (Klk2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Pro19~Pro261. Applications: WB, IHC, ICC, IP. Target Synonyms: Kallikrein 2 (KLK2). Accession Number: P20151. Gene ID: 3817. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9309