Learn More
Biomatik Corporation Hemoglobin Alpha 1 (Hba1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU23588
Encompass_Preferred
Hemoglobin Alpha 1 (Hba1) Polyclonal Antibody is a Bovine (Bos taurus) Polyclonal Antibody to Hemoglobin Alpha 1 (HBa1). The Hemoglobin Alpha 1 (Hba1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Bovine (Bos taurus). Immunogen: Val2~Arg142. Applications: WB, IHC, ICC, IP. Target Synonyms: Hemoglobin Alpha 1 (HBa1). Accession Number: P01966. Gene ID: 100140149. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9312