Learn More
Biomatik Corporation Quinoid Dihydropteridine Reductase (Qdpr) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU23898
Encompass_Preferred
Quinoid Dihydropteridine Reductase (Qdpr) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Quinoid Dihydropteridine Reductase (QDPR). The Quinoid Dihydropteridine Reductase (Qdpr) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Phe244. Applications: WB, IHC, ICC, IP. Target Synonyms: Quinoid Dihydropteridine Reductase (QDPR). Accession Number: P09417. Gene ID: 5860. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9324