Learn More
Biomatik Corporation Collagen Type Vi Alpha 3 (Col6A3) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU24610
Encompass_Preferred
Collagen Type Vi Alpha 3 (Col6A3) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Collagen Type VI Alpha 3 (COL6a3). The Collagen Type Vi Alpha 3 (Col6A3) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Asp2009~Leu2205. Applications: WB, IHC, ICC, IP. Target Synonyms: Collagen Type VI Alpha 3 (COL6a3). Accession Number: J3QQ16. Gene ID: 12835. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9353