Learn More
Biomatik Corporation Collagen Type Iv Alpha 5 (Col4A5) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU24620
Encompass_Preferred
Collagen Type Iv Alpha 5 (Col4A5) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Collagen Type IV Alpha 5 (COL4a5). The Collagen Type Iv Alpha 5 (Col4A5) Polyclonal Antibody has been validated for the following applications: WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Gly1461~Thr1685. Applications: WB. Target Synonyms: Collagen Type IV Alpha 5 (COL4a5). Accession Number: P29400. Gene ID: 1287. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9357