Learn More
Biomatik Corporation Sp140 Nuclear Body Protein (Sp140) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU21241
Encompass_Preferred
Sp140 Nuclear Body Protein (Sp140) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Sp140 Nuclear Body Protein (SP140). The Sp140 Nuclear Body Protein (Sp140) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Gly443~Met655. Applications: WB, IHC, ICC, IP. Target Synonyms: Sp140 Nuclear Body Protein (SP140). Accession Number: Q13342. Gene ID: 11262. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9199