Learn More
Biomatik Corporation Hornerin (Hrnr) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU21840
Encompass_Preferred
Hornerin (Hrnr) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Hornerin (HRNR). The Hornerin (Hrnr) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Glu2685~Gln2850. Applications: WB, IHC, ICC, IP. Target Synonyms: Hornerin (HRNR). Accession Number: Q86YZ3. Gene ID: 388697. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9222