Learn More
Biomatik Corporation Sulfatase 2 (Sulf2) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU22062
Encompass_Preferred
Sulfatase 2 (Sulf2) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Sulfatase 2 (SULF2). The Sulfatase 2 (Sulf2) Polyclonal Antibody has been validated for the following applications: WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Phe337~Arg649. Applications: WB. Target Synonyms: Sulfatase 2 (SULF2). Accession Number: Q8IWU5. Gene ID: 55959. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9237