Learn More
Biomatik Corporation Uracil Dna Glycosylase (Ung) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU22129
Encompass_Preferred
Uracil Dna Glycosylase (Ung) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Uracil DNA Glycosylase (UNG). The Uracil Dna Glycosylase (Ung) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Leu313. Applications: WB, IHC, ICC, IP. Target Synonyms: Uracil DNA Glycosylase (UNG). Accession Number: P13051. Gene ID: 7374. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9241