Learn More
Biomatik Corporation Lactate Dehydrogenase A (Ldha) Polyclonal Antibody
Catalog No. 502289434
Encompass_Preferred
Lactate Dehydrogenase A (Ldha) Polyclonal Antibody is a Pig (Sus scrofa; Porcine) Polyclonal Antibody to Lactate Dehydrogenase A (LDHA). The Lactate Dehydrogenase A (Ldha) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Pig (Sus scrofa; Porcine). Immunogen: Met1~Phe332. Applications: WB, IHC, ICC, IP. Target Synonyms: Lactate Dehydrogenase A (LDHA). Accession Number: P00339. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-228-9434 Supplier Biomatik Corporation Supplier No. CAU26054
Add to Cart
Add to Cart
Lactate Dehydrogenase A (Ldha) Polyclonal Antibody is a Pig (Sus scrofa; Porcine) Polyclonal Antibody to Lactate Dehydrogenase A (LDHA). The Lactate Dehydrogenase A (Ldha) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Pig (Sus scrofa; Porcine). Immunogen: Met1~Phe332. Applications: WB, IHC, ICC, IP. Target Synonyms: Lactate Dehydrogenase A (LDHA). Accession Number: P00339. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.