Learn More
Biomatik Corporation Galactosylceramidase (Galc) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU22198
Encompass_Preferred
Galactosylceramidase (Galc) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Galactosylceramidase (GALC). The Galactosylceramidase (Galc) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Asp375~Asn535. Applications: WB, IHC, ICC, IP. Target Synonyms: Galactosylceramidase (GALC). Accession Number: P54803. Gene ID: 2581. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9245