Learn More
Biomatik Corporation Fructosamine-3-Kinase (Fn3K) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU21800
Encompass_Preferred
Fructosamine-3-Kinase (Fn3K) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Fructosamine-3-Kinase (FN3K). The Fructosamine-3-Kinase (Fn3K) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Lys309. Applications: WB, IHC, ICC, IP. Target Synonyms: Fructosamine-3-Kinase (FN3K). Accession Number: Q9H479. Gene ID: 64122. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9219