Learn More
Biomatik Corporation Prolactin (Prl) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26971
Encompass_Preferred
Prolactin (Prl) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Prolactin (PRL). The Prolactin (Prl) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Leu29~Cys227. Applications: WB, IHC, ICC, IP. Target Synonyms: Prolactin (PRL). Accession Number: Q5THQ0. Gene ID: 5617. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9490