Learn More
Biomatik Corporation Uroplakin 3A (Upk3A) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU22446
Encompass_Preferred
Uroplakin 3A (Upk3A) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Uroplakin 3A (UPK3A). The Uroplakin 3A (Upk3A) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Cys15~Thr212. Applications: WB, IHC, ICC, IP. Target Synonyms: Uroplakin 3A (UPK3A). Accession Number: D3ZZ76. Gene ID: 315190. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9260