Learn More
Biomatik Corporation Glutamate Receptor, Ionotropic, Ampa 3 (Gria3) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU22663
Encompass_Preferred
Glutamate Receptor, Ionotropic, Ampa 3 (Gria3) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Glutamate Receptor, Ionotropic, AMPA 3 (GRIA3). The Glutamate Receptor, Ionotropic, Ampa 3 (Gria3) Polyclonal Antibody has been validated for the following applications: WB, ICC, IHC-P, IHC-F, ELISA. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Thr649~Asn823. Applications: WB, ICC, IHC-P, IHC-F, ELISA. Target Synonyms: Glutamate Receptor, Ionotropic, AMPA 3 (GRIA3). Accession Number: P42263. Gene ID: 2892. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9269