Learn More
Biomatik Corporation Microfibrillar Associated Protein 2 (Mfap2) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU24043
Encompass_Preferred
Microfibrillar Associated Protein 2 (Mfap2) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Microfibrillar Associated Protein 2 (MFAP2). The Microfibrillar Associated Protein 2 (Mfap2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Leu6~Val162. Applications: WB, IHC, ICC, IP. Target Synonyms: Microfibrillar Associated Protein 2 (MFAP2). Accession Number: P55001. Gene ID: 4237. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9330