Learn More
Biomatik Corporation Cytidine Deaminase (Cda) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU24358
Encompass_Preferred
Cytidine Deaminase (Cda) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Cytidine Deaminase (CDA). The Cytidine Deaminase (Cda) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ala2~Gln146. Applications: WB, IHC, ICC, IP. Target Synonyms: Cytidine Deaminase (CDA). Accession Number: P32320. Gene ID: 978. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9342