Learn More
Biomatik Corporation Complement Component 5A (C5A) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27998
Encompass_Preferred
Complement Component 5A (C5A) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Complement Component 5a (C5a). The Complement Component 5A (C5A) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Asn679~Arg755. Applications: WB, IHC, ICC, IP. Target Synonyms: Complement Component 5a (C5a). Accession Number: P06684. Gene ID: 15139. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9533