Learn More
Biomatik Corporation Brevican (Bcan) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU24580
Encompass_Preferred
Brevican (Bcan) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Brevican (BCAN). The Brevican (Bcan) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Asp23~Arg353. Applications: WB, IHC. Target Synonyms: Brevican (BCAN). Accession Number: Q61361. Gene ID: 12032. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9352