Learn More
Biomatik Corporation Interferon Alpha 4 (Ifna4) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU28406
Encompass_Preferred
Interferon Alpha 4 (Ifna4) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Interferon Alpha 4 (IFNa4). The Interferon Alpha 4 (Ifna4) Polyclonal Antibody has been validated for the following applications: WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ala10~Ala163. Applications: WB. Target Synonyms: Interferon Alpha 4 (IFNa4). Accession Number: P05014. Gene ID: 3441. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9544