Learn More
Biomatik Corporation Interleukin 33 (Il33) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU24925
Encompass_Preferred
Interleukin 33 (Il33) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Interleukin 33 (IL33). The Interleukin 33 (Il33) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Ser109~Ile266. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 33 (IL33). Accession Number: Q8BVZ5. Gene ID: 77125. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9369