Learn More
Biomatik Corporation Ficolin 2 (Fcn2) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU25088
Encompass_Preferred
Ficolin 2 (Fcn2) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Ficolin 2 (FCN2). The Ficolin 2 (Fcn2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Gln97~Ser254. Applications: WB, IHC, ICC, IP. Target Synonyms: Ficolin 2 (FCN2). Accession Number: O70497. Gene ID: 14134. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9378