Learn More
Biomatik Corporation Cyclin Dependent Kinase 2 (Cdk2) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU25117
Encompass_Preferred
Cyclin Dependent Kinase 2 (Cdk2) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Cyclin Dependent Kinase 2 (CDK2). The Cyclin Dependent Kinase 2 (Cdk2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ser94~Thr198. Applications: WB, IHC, ICC, IP. Target Synonyms: Cyclin Dependent Kinase 2 (CDK2). Accession Number: P24941. Gene ID: 1017. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9382