Learn More
Biomatik Corporation Interferon Regulatory Factor 1 (Irf1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU25711
Encompass_Preferred
Interferon Regulatory Factor 1 (Irf1) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Interferon Regulatory Factor 1 (IRF1). The Interferon Regulatory Factor 1 (Irf1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: His44~Pro319. Applications: WB, IHC, ICC, IP. Target Synonyms: Interferon Regulatory Factor 1 (IRF1). Accession Number: P15314. Gene ID: 16362. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9409